Learn More
Aviva Systems Biology Corp STAT1 Antibody - N-terminal region (ARP33373_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33373_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STAT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STAT1. Alternate names: DKFZp686B04100, ISGF-3, STAT91, CANDF7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54985