Learn More
Abcam STAT1 (pY701) ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1264561X96TESTS
Encompass_Preferred
STAT1 (pY701) ELISA Kit is a Semi-quantitative ELISA kit for the measurement of STAT1 (pY701) in Mouse, Human in Cell Lysate samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1545256