Learn More
Abcam STAT1 (pY701) + total STAT1 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1264571X96TESTS
Encompass_Preferred
STAT1 (pY701) + total STAT1 ELISA Kit is a Semi-quantitative ELISA kit for the measurement of STAT1 (pY701) + total STAT1 in Mouse, Human in Cell Lysate samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1037525