Learn More
Aviva Systems Biology Corp STAT3 Antibody - middle region (ARP30317_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30317_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STAT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human STAT3. Alternate names: APRF, FLJ20882, MGC16063, HIESApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54154