Learn More
Santa Cruz Biotechnology Anti-Stat3 Antibody (F-2), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC8019
Encompass
Anti-Stat3 Antibody (F-2) is recommended for detection of Stat3 of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA; also reactive with additional species, including and equine, bovine and porcine.
Catalog No. NC0902706