Learn More
Selleck Chemical LLC STAT3-IN-1 5mg
SDP
Supplier:  Selleck Chemical LLC S08185MG
Encompass_Preferred
STAT3-IN-1 (compound 7d) is an excellent, selective and orally active inhibitor of STAT3 with IC50 of 1.82 ?M and 2.14 ?M in HT29 and MDA-MB 231 cells, respectively. STAT3-IN-1 induces apoptosis of tumor cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3047