Learn More
Medchemexpress LLC HY-100753 5mg Medchemexpress, STAT3-IN-1 CAS:2059952-75-7 Purity:>98%
SDP
Catalog No. 502029472
Encompass_Preferred
Medchemexpress, HY-100753 5mg STAT3-IN-1 CAS:2059952-75-7 STAT3-IN-1 (compound 7d) is an excellent, selective and orally active STAT3 inhibitor, with IC50 values of 1.82 μM and 2.14 μM in HT29 and MDA-MB 231 cells, respectively. STAT3-IN-1 (compound 7d) induces tumor apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9472 Supplier Medchemexpress LLC Supplier No. HY1007535MG
Medchemexpress, HY-100753 5mg STAT3-IN-1 CAS:2059952-75-7 STAT3-IN-1 (compound 7d) is an excellent, selective and orally active STAT3 inhibitor, with IC50 values of 1.82 μM and 2.14 μM in HT29 and MDA-MB 231 cells, respectively. STAT3-IN-1 (compound 7d) induces tumor apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.