Learn More
Cayman Chemical STAT3 InhIbItr 4m 10mg
SDP
Catalog No. 502915479
Encompass_Preferred
A STAT3 inhibitor; inhibits the phosphorylation of STAT3 and decreases levels of survivin and Mcl-1 in HCT116 cells at 0.75 and 1 µM; inhibits the proliferation of A549, HCT116, and HepG2 cells (IC50s = 0.93, 0.61, and 1.79 µM, respectively); induces apoptosis and cell cycle arrest at both the S and G2/M phases in HCT116 cells at 5 µM; reduces the viability of human CCOs from 1-10 µM
Catalog No. 50-291-5479 Supplier Cayman Chemical Supplier No. 3735210 MG
Add to Cart
Add to Cart
A STAT3 inhibitor; inhibits the phosphorylation of STAT3 and decreases levels of survivin and Mcl-1 in HCT116 cells at 0.75 and 1 µM; inhibits the proliferation of A549, HCT116, and HepG2 cells (IC50s = 0.93, 0.61, and 1.79 µM, respectively); induces apoptosis and cell cycle arrest at both the S and G2/M phases in HCT116 cells at 5 µM; reduces the viability of human CCOs from 1-10 µM