Learn More
Enzo Life Sciences STAT3 (pTyr705) monoclonal antibody (9E12) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804180C100
Encompass_Preferred
Clone: 9E12. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of STAT3 phosphorylated at Tyr705 conjugated to KLH. Species reactivity: Human, Mouse, Dog. Specificity: Recognizes the -AAPpYLKT-motif of activated STAT3 phosphorylated at Tyr705. Applications: ELISA, ICC, IHC, IP, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9818