Learn More
Enzo Life Sciences STAT3 (pSer727) monoclonal antibody (23G5) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905672100
Encompass_Preferred
Clone: 23G5. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of Stat3 phosphorylated at Ser727. UniProt ID: P40763. Species reactivity: Human, Mouse, Rat, Dog. Applications: ELISA, IHC, WB. Recommended Dilutions: ELISA (0.1µg/ml) Immunohistochemistry (1:50) Western Blot (0.5µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-6987