Learn More
Enzo Life Sciences STAT3 (pTyr705) polyclonal antibody (20 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA3280020
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of human STAT3 phosphorylated at Tyr705. Species reactivity: Human, Mouse, Rat. Specificity: Recognizes STAT3 phosphorylated at Tyr705. Applications: WB. Formulation: Liquid. In PBS containing BSA and sodium azide.
Catalog No. 50-201-1800