Learn More
Enzo Life Sciences STAT3 (pTyr705) polyclonal antibody (25µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPTF022C
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Stat3 phosphorylated at Tyr705. UniProt ID: P40763. Source: Purified from rabbit serum. Species reactivity: Human. Creossreactivity: Weakly cross-reacts with mouse, rat, bovine and pig Stat3. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7303