Learn More
BPS Bioscience Inc STAT3 Reporter Kit (STAT3 Signaling Pathway) | 500 Reactions
SDP
Catalog No. NC1754228
Encompass
The STAT3 Reporter kit is designed for monitoring the activity of the STAT3 signaling pathway in cultured cells. The kit contains transfection-ready STAT3 luciferase reporter vector. This reporter contains a firefly luciferase gene under the control of STAT3-responsive element located upstream of a minimal promoter. The STAT3 reporter is premixed with constitutively expressing Renilla luciferase vector, which serves as an internal control for transfection efficiency. The kit also includes a non-inducible firefly luciferase vector premixed with constitutively-expressing Renilla luciferase vector as a negative control. The non-inducible luciferase vector contains the firefly luciferase gene under the control of a minimal promoter, without any additional response elements. The negative control is critical to determining pathway-specific effects and background luciferase activity.
Catalog No. NC1754228 Supplier BPS Bioscience Inc Supplier No. 79730
The STAT3 Reporter kit is designed for monitoring the activity of the STAT3 signaling pathway in cultured cells. The kit contains transfection-ready STAT3 luciferase reporter vector. This reporter contains a firefly luciferase gene under the control of STAT3-responsive element located upstream of a minimal promoter. The STAT3 reporter is premixed with constitutively expressing Renilla luciferase vector, which serves as an internal control for transfection efficiency. The kit also includes a non-inducible firefly luciferase vector premixed with constitutively-expressing Renilla luciferase vector as a negative control. The non-inducible luciferase vector contains the firefly luciferase gene under the control of a minimal promoter, without any additional response elements. The negative control is critical to determining pathway-specific effects and background luciferase activity.