Learn More
Cayman Chemical STAT3aPhospho-Tyr705 Rab 100uL
SDP
Catalog No. 502947824
Encompass_Preferred
An inhibitor of the protein-protein interaction between PD-1 and PDL1 (IC50 1.4 nM in a HTRF assay) increases the activation of Jurkat cells expressing PD-1 in co-culture with CHO cells expressing PD-L1 (EC50 276 nM in a reporter assay)
Catalog No. 50-294-7824 Supplier Cayman Chemical Supplier No. 32212100 UL
An inhibitor of the protein-protein interaction between PD-1 and PDL1 (IC50 1.4 nM in a HTRF assay) increases the activation of Jurkat cells expressing PD-1 in co-culture with CHO cells expressing PD-L1 (EC50 276 nM in a reporter assay)