Learn More
Medchemexpress LLC HY-101853 5mg Medchemexpress, STAT5-IN-1 CAS:285986-31-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1018535MG
Encompass_Preferred
Medchemexpress, HY-101853 5mg STAT5-IN-1 CAS:285986-31-4 STAT5-IN-1 is a STAT5 inhibitor with an IC50 of 47 μM for STAT5β isoform. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7888