Learn More
Cayman Chemical STAT5 InhIbItr 5mg
SDP
Catalog No. 502973039
Encompass_Preferred
A cell-permeable, nonpetidic nicotinoyl hydrazone that selectively suppresses STAT5 by targeting its SH2 domain (IC50 = 47 µM against STAT5β SH2 domain EPO peptide binding activity)
Catalog No. 50-297-3039 Supplier Cayman Chemical Supplier No. 157845 MG
A cell-permeable, nonpetidic nicotinoyl hydrazone that selectively suppresses STAT5 by targeting its SH2 domain (IC50 = 47 µM against STAT5β SH2 domain EPO peptide binding activity)