Learn More
Bioss Stat5 - Tyr-694 Phosphospecif
SDP
Catalog No. 5000065839
Encompass_Preferred
Stat5 (Tyr-694) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M148Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65839 Supplier Bioss Supplier No. BSM70688M
Stat5 (Tyr-694) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M148Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol