Learn More
Aviva Systems Biology Corp STAT5A Antibody - C-terminal region (ARP38256_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38256_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STAT5A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human STAT5A. Alternate names: MGF, STAT5ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-56583