Learn More
Aviva Systems Biology Corp STAT5B Antibody - N-terminal region (ARP39000_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39000_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STAT5B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STAT5B. Alternate names: STAT5ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56883