Learn More
Aviva Systems Biology Corp STAT6 Antibody - C-terminal region (ARP38257_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38257_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STAT6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human STAT6. Alternate names: D12S1644, IL-4-STAT, STAT6B, STAT6CApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56584