Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Signal transducer and activator of transcription 6 (STAT6), human, phosphorylated | >90.0% | 10UG
SDP

Catalog No. 5000174112
Encompass_Preferred

Phosphorylated human STAT6 is a recombinant, N-terminal 6xHis-tagged protein (536 amino acids) supplied as a purified phosphorylated form for use in biochemical and signaling research. Expressed in E. coli, it is intended for studies of IL-4/IL-13-mediated STAT6 activation, in vitro assays, and antibody validation.

  • Recombinant phosphorylated human STAT6 for signaling studies.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Sequence length 536 amino acids; molecular weight ~61.7 kDa.
  • Purity >90% by reducing SDS-PAGE.
  • Expressed in E. coli for consistent batch production.
  • Store at -80°C; avoid repeated freeze-thaw cycles.

Catalog No. 50-001-74112 Supplier Medchemexpress LLC Supplier No. HYP70283510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphorylated human STAT6 is a recombinant, N-terminal 6xHis-tagged protein (536 amino acids) supplied as a purified phosphorylated form for use in biochemical and signaling research. Expressed in E. coli, it is intended for studies of IL-4/IL-13-mediated STAT6 activation, in vitro assays, and antibody validation.

  • Recombinant phosphorylated human STAT6 for signaling studies.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Sequence length 536 amino acids; molecular weight ~61.7 kDa.
  • Purity >90% by reducing SDS-PAGE.
  • Expressed in E. coli for consistent batch production.
  • Store at -80°C; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.