Learn More
Medchemexpress LLC STAT6-IN-2 INHIBITOR 5MG
SDP
Supplier:  Medchemexpress LLC HY1539915MG
Encompass_Preferred
STAT6-IN-2 (Comp R-84) is an inhibitor of STAT6. STAT6-IN-2 inhibits the secretion of chemokine eliciting eosinophil infiltration eotaxin-3. STAT6-IN-2 can be used for immune disease research.
Catalog No. NC2893298