Learn More
Desco™ Statfree J™ Dissipative Shock Resistant Foam, Rubber Roll
SDP
Catalog No. 18688130
Click to view available options
Dimensions (L x W):
40 x 2 ft.
Catalog No. Dimensions (L x W)
18-688-130 40 x 2 ft.
1 options
Catalog No. 18-688-130 Supplier Desco™ Supplier No. 45000
Rubber Roll designed to provide a cushioned surface that PCB and other products will not slide off of.
  • Non-skid foam nitrile butadiene rubber
  • Dissipative Worksurface - 10E6 - 10E8 Ohms RTT
  • Controls charge removal from ESD susceptible items; meets ANSI/ESD S20.20 worksurface required limit
  • Volume Resistance 10E6 - 10E7 Ohms per ANSI/ESD STM11.12 allows the mat to electrically couple to the grounded ESD mat it is being placed on without a separate ground cord for the Disposable ESD Mat
  • Lead-free RoHS compliant
  • Limited Lifetime Warranty
  • Electricals guaranteed for the life of the mat
Specifications
Dimensions (L x W) 40 x 2 ft.
Material Foam Rubber
Color Black