Learn More
Aviva Systems Biology Corp STATH Antibody - middle region (ARP42102_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42102_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STATH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human STATH. Alternate names: STRApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-58108