Learn More
ACROBiosystems Recombinant Protein;Human Stathmin 1 / STMN1 Protein;E.coli;1MG;ST1,STMN1,Stathmin,Metablastin,Oncoprotein 18,Op18,C1orf215,LAP18,OP18,pp19,pp17,Protein Pr22
Supplier:  ACROBiosystems ST1H51491MG
Encompass_Preferred
Recombinant Protein;Human Stathmin 1 / STMN1 Protein;1MG;Human Stathmin 1, His Tag (ST1-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 149 (Accession # NP_981946).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.1 kDa. The protein migrates as 17-20 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8833