Learn More
MSC Bosch Anti-Static Bend Nozzle, Plastic, Anti-Static, 35 mm and 1-1/2 in
Catalog No. 502837223
Encompass
The Bosch VX100 is an anti-static bend nozzle constructed from rugged plastic. It is designed for professional use, fitting 35 mm wands and hoses, as well as most 1-1/2 inch wands and hoses. This nozzle is a standard component with Bosch VAC series dust extractors, providing efficient and anti-static operation in various applications.
  • Anti-static bend nozzle
  • Fits 35 mm wands and hoses
  • Fits most 1-1/2 in wands and hoses
  • Standard item with VAC series dust extractors
  • Made in Italy
  • Rugged construction
  • Clicks onto VH series hoses
  • Friction fits PowerGrip adapter
Catalog No. 50-283-7223 Supplier MSC Supplier No. 47824107
The Bosch VX100 is an anti-static bend nozzle constructed from rugged plastic. It is designed for professional use, fitting 35 mm wands and hoses, as well as most 1-1/2 inch wands and hoses. This nozzle is a standard component with Bosch VAC series dust extractors, providing efficient and anti-static operation in various applications.
  • Anti-static bend nozzle
  • Fits 35 mm wands and hoses
  • Fits most 1-1/2 in wands and hoses
  • Standard item with VAC series dust extractors
  • Made in Italy
  • Rugged construction
  • Clicks onto VH series hoses
  • Friction fits PowerGrip adapter