Learn More
MSC Guardair Static Conductive Air Hose Assembly, 3/4 in ID, 50 ft
Catalog No. 502837385
Encompass
This static conductive air hose assembly is designed for supplying compressed air in applications where static electricity is a concern. It features reinforced rubber construction for ozone resistance and durability. The assembly includes swivel male NPT connectors on both ends for easy installation and is manufactured to industry standards.
  • End to end resistance of less than 10^6 Ohms.
  • Maximum pressure: 200 psi.
  • Swivel male NPT fittings on both ends for ease of installation.
  • Reinforced rubber provides excellent ozone resistance and long-term durability.
  • Manufactured and tested to industry standards.
  • Ideal for supplying compressed air to vacuums in applications where static electricity is a concern.
Catalog No. 50-283-7385 Supplier MSC Supplier No. 64355670
This static conductive air hose assembly is designed for supplying compressed air in applications where static electricity is a concern. It features reinforced rubber construction for ozone resistance and durability. The assembly includes swivel male NPT connectors on both ends for easy installation and is manufactured to industry standards.
  • End to end resistance of less than 10^6 Ohms.
  • Maximum pressure: 200 psi.
  • Swivel male NPT fittings on both ends for ease of installation.
  • Reinforced rubber provides excellent ozone resistance and long-term durability.
  • Manufactured and tested to industry standards.
  • Ideal for supplying compressed air to vacuums in applications where static electricity is a concern.