Learn More
MSC Guardair Multipurpose Air Hose, Static Conductive, Swivel NPT, 12 ft, 3/8 in ID
Catalog No. 502838088
Encompass
Guardair Multipurpose Air Hose is designed for supplying compressed air in static-sensitive environments. This heavy-duty rubber hose provides excellent ozone resistance and durability, and includes swivel male NPT connectors for easy installation.
  • Ideal for supplying compressed air in applications where static electricity is of concern.
  • Reinforced rubber construction offers excellent ozone resistance and durability.
  • Manufactured and tested to industry standards.
  • Includes swivel male NPT connectors on both ends for ease of installation.
Catalog No. 50-283-8088 Supplier MSC Supplier No. 64355928
Guardair Multipurpose Air Hose is designed for supplying compressed air in static-sensitive environments. This heavy-duty rubber hose provides excellent ozone resistance and durability, and includes swivel male NPT connectors for easy installation.
  • Ideal for supplying compressed air in applications where static electricity is of concern.
  • Reinforced rubber construction offers excellent ozone resistance and durability.
  • Manufactured and tested to industry standards.
  • Includes swivel male NPT connectors on both ends for ease of installation.