Learn More
MSC Guardair Multipurpose Air Hose, 3/8 inch ID, 50 ft. Length, Static Conductive, Swivel Ends
Catalog No. 502838795
Encompass
Guardair multipurpose air hoses are designed for applications where static electricity is a concern. Constructed from heavy-duty reinforced rubber, these hoses offer excellent ozone resistance and durability. They feature swivel male NPT connectors on both ends for easy installation. Ideal for supplying compressed air to vacuums, these hoses are manufactured and tested to industry standards.
  • Ideal for supplying compressed air to vacuums in applications where static electricity is of concern.
  • Reinforced rubber provides excellent ozone resistance and long-term durability.
  • Manufactured and tested to industry standards.
  • Includes swivel male NPT connectors on both ends for ease of installation.
Catalog No. 50-283-8795 Supplier MSC Supplier No. 64355977
Guardair multipurpose air hoses are designed for applications where static electricity is a concern. Constructed from heavy-duty reinforced rubber, these hoses offer excellent ozone resistance and durability. They feature swivel male NPT connectors on both ends for easy installation. Ideal for supplying compressed air to vacuums, these hoses are manufactured and tested to industry standards.
  • Ideal for supplying compressed air to vacuums in applications where static electricity is of concern.
  • Reinforced rubber provides excellent ozone resistance and long-term durability.
  • Manufactured and tested to industry standards.
  • Includes swivel male NPT connectors on both ends for ease of installation.