Learn More
Electron Microscopy Sciences Static Control Adjustable Wrist Band
SDP
Catalog No. 50996261
Encompass_Preferred
This Hook and Loop Wrist Strap is designed for those whom need to be static free in the working area. This Hook and Loop wrist trap has a quick and easy adjustment. With a 1" wide conductive material that is sewn in and has a 1M ohms resistor.
Catalog No. 50-996-261 Supplier Electron Microscopy Sciences Supplier No. 6062212
This Hook and Loop Wrist Strap is designed for those whom need to be static free in the working area. This Hook and Loop wrist trap has a quick and easy adjustment. With a 1" wide conductive material that is sewn in and has a 1M ohms resistor.