Learn More
Electron Microscopy Sciences Static Control Adjustable Wrist Band, 6' Coil Cord
SDP
Catalog No. 50996260
Encompass_Preferred
This Hook and Loop Wrist Strap is designed for those whom need to be static free in the working area. This Hook and Loop wrist trap has a quick and easy adjustment. With a 1" wide conductive material that is sewn in and has a 1M ohms resistor.
This item is not returnable. View return policy
Catalog No. 50-996-260 Supplier Electron Microscopy Sciences Supplier No. 6062206
This Hook and Loop Wrist Strap is designed for those whom need to be static free in the working area. This Hook and Loop wrist trap has a quick and easy adjustment. With a 1" wide conductive material that is sewn in and has a 1M ohms resistor.