Learn More
MSC ProTeam Static-Dissipating Hose, 1.5 in, 48 in, Cuffed, 1 ea
Catalog No. 502866478
Encompass
This static-dissipating hose is a replacement accessory designed for ProTeam backpack vacuums. It features static-dissipating construction to prevent static charge buildup and measures 1.5 in. in diameter by 48 in. in length, complete with pre-fitted cuffs on both ends.
  • Reduces static charge buildup, ideal for environments with sensitive electronics or flammable materials
  • Equipped with cuffs on both ends for secure attachment to vacuum body and wand connection
  • Compatible with various backpack vacuum models, including GoFit 3, GoFit 10, and GoFree Flex Pro II
Catalog No. 50-286-6478 Supplier MSC Supplier No. 59276485
This static-dissipating hose is a replacement accessory designed for ProTeam backpack vacuums. It features static-dissipating construction to prevent static charge buildup and measures 1.5 in. in diameter by 48 in. in length, complete with pre-fitted cuffs on both ends.
  • Reduces static charge buildup, ideal for environments with sensitive electronics or flammable materials
  • Equipped with cuffs on both ends for secure attachment to vacuum body and wand connection
  • Compatible with various backpack vacuum models, including GoFit 3, GoFit 10, and GoFree Flex Pro II