Learn More
Innovive Inc Static lid, low-profile ventilation area
Catalog No. NC1555788
Encompass
A low-profile ventilation area static mouse lid with large dual Uniblend filter areas and metal grates for use in emergencies. It fits into the Innovive standard IVC rack in the event of a power failure.
Catalog No. NC1555788 Supplier Innovive Inc Supplier No. MSX2
A low-profile ventilation area static mouse lid with large dual Uniblend filter areas and metal grates for use in emergencies. It fits into the Innovive standard IVC rack in the event of a power failure.