Learn More
Electron Microscopy Sciences DiATOME Static Line Ionizer
SDP
Catalog No. 501900195
Encompass
The Static Line II ionizer is an antistatic device which emits negative and positive ions, neutralizing the electrostatic charging in the cryochamber. The sections no longer stick to the knife edge, or bunch up one on top of the other, but float in a nice ribbon over the knife surface. The pickup of the sections can be done easily. We recommend the use of the Static Line II ionizer for trimming, sectioning and section collection.
Catalog No. 50-190-0195 Supplier Electron Microscopy Sciences Supplier No. SLI
The Static Line II ionizer is an antistatic device which emits negative and positive ions, neutralizing the electrostatic charging in the cryochamber. The sections no longer stick to the knife edge, or bunch up one on top of the other, but float in a nice ribbon over the knife surface. The pickup of the sections can be done easily. We recommend the use of the Static Line II ionizer for trimming, sectioning and section collection.