Learn More
Rice Lake Weighing System Ionizer,1 x 3-inch NRD Staticmaster 1 x 3-inch Ionizing Unit
Catalog No. NC9486091
Encompass_Preferred
Eliminate unwanted, damaging electrostatic discharge with NRD ionizing strips. Ionizer, Rice Lake Weighing Systems 1 x 3-inch NRD Staticmaster 1 x 3-inch Ionizing Unit
Catalog No. NC9486091 Supplier Rice Lake Weighing System Supplier No. 76676
Eliminate unwanted, damaging electrostatic discharge with NRD ionizing strips. Ionizer, Rice Lake Weighing Systems 1 x 3-inch NRD Staticmaster 1 x 3-inch Ionizing Unit