Learn More
Electron Microscopy Sciences Static Master Static Master Model 1C200
SDP
Catalog No. 5030514
Encompass_Preferred
Eliminate static electric charges, and remove dust, lint and other foreign particles in one quick operation. StaticMaster contains Polonium210 element' which emits low energy alpha particles. Alpha particles are non-penetrating, stopped completely by a thin sheet of paper or a few inches of air. By ionizing the air in the immediate vicinity of the film, lens, etc., a conductive path is provided through which the static change is safely and effectively grounded. This 1" model cleans negatives, mounted or u nmounted transparencies, cameras, and smaller enlarging lenses.
This item is not returnable. View return policy
Catalog No. 50-305-14 Supplier Electron Microscopy Sciences Supplier No. 606131C
Eliminate static electric charges, and remove dust, lint and other foreign particles in one quick operation. StaticMaster contains Polonium210 element' which emits low energy alpha particles. Alpha particles are non-penetrating, stopped completely by a thin sheet of paper or a few inches of air. By ionizing the air in the immediate vicinity of the film, lens, etc., a conductive path is provided through which the static change is safely and effectively grounded. This 1" model cleans negatives, mounted or u nmounted transparencies, cameras, and smaller enlarging lenses.