Learn More
Electron Microscopy Sciences StaticMaster 3C500 w/retractable brush
SDP
Catalog No. 5030512
Encompass_Preferred
Eliminate static electric charges, and remove dust, lint and other foreign particles in one quick operation. StaticMaster contains Polonium210 element' which emits low energy alpha particles. Alpha particles are non-penetrating, stopped completely by a thin sheet of paper or a few inches of air. By ionizing the air in the immediate vicinity of the film, lens, etc., a conductive path is provided through which the static change is safely and effectively grounded. The 3" model cleans large areas quickly and effectively. Its brush is retractable to just a desired length.
Catalog No. 50-305-12 Supplier Electron Microscopy Sciences Supplier No. 606133C
Eliminate static electric charges, and remove dust, lint and other foreign particles in one quick operation. StaticMaster contains Polonium210 element' which emits low energy alpha particles. Alpha particles are non-penetrating, stopped completely by a thin sheet of paper or a few inches of air. By ionizing the air in the immediate vicinity of the film, lens, etc., a conductive path is provided through which the static change is safely and effectively grounded. The 3" model cleans large areas quickly and effectively. Its brush is retractable to just a desired length.