Learn More
Thermo Scientific 350 µL Static Mixer, Vanquish H/F Pumps
Supplier:  Thermo Scientific 6044.5310
Encompass
For Use With (Equipment): Vanquish Binary Pump H: Optional, Vanquish Binary Pump F: Optional, Vanquish Quaternary Pump F: Default, Vanquish Dual Pump F: Default | Includes Static mixer: 350 µL
Catalog No. NC1611914