Learn More
Uline Static Shielding Bags S-1313, Translucent Metallic, 100 per carton
Catalog No. NC2112593
Encompass
Prevent incoming static charges from damaging ESD-sensitive products, 3 mil translucent metallic bags for easy content identification, Metal Faraday cage layer shields products from electric energy inside and prevents static buildup, 4-layer protection guards against charges inside and out, MEETS THE REQUIREMENTS OF MIL-STD-3010 4046 EIA 541 EIA 625 AND ANSIESD S2020
This item is not returnable. View return policy
Catalog No. NC2112593 Supplier Uline Supplier No. S1313
Prevent incoming static charges from damaging ESD-sensitive products, 3 mil translucent metallic bags for easy content identification, Metal Faraday cage layer shields products from electric energy inside and prevents static buildup, 4-layer protection guards against charges inside and out, MEETS THE REQUIREMENTS OF MIL-STD-3010 4046 EIA 541 EIA 625 AND ANSIESD S2020