Learn More
Static Solutions™ OHM-Stat™ CT-8900 Combination Tester/Data Logger
Catalog No. 191306021
Click to view available options
Includes:
Combination Tester/Data Logger
Combo Tester w/ Footplate, Stand, Barxode or Mag Stripe Reader
Combo Tester w/ Footplate, Stand, HID Proximity Reader
Combo Tester w/ Software Only
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-130-6021 Combo Tester w/ Footplate, Stand, HID Proximity Reader
19-130-6023 Combo Tester w/ Software Only
19-130-6018 Combination Tester/Data Logger
19-130-6020 Combo Tester w/ Footplate, Stand, Barxode or Mag Stripe Reader
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-130-6021 Supplier Static Solutions™ Supplier No. CT8960
Includes
NIST certificate.
No more log books—tests foot grounder and wrist strap in three seconds
Measures the electrical resistance of each foot and wrist strap simultaneously and individually with one push of a button. Offers adjustable limits, exact resistivity readings, unique report generation groups, versatile networking options, and an expandable employee log of 30,000. Made in USA.
  • Increases worker productivity, reduces defects, and saves administrative time
  • Accepts mag stripe, barcode, Motorola™, HID™, and Casi-Rusco™ proximity ID cards as well as keyboard entry
  • Included ESD Protection Test Log software generates e-mail reports to multiple recipients
  • Logs are easily imported into spreadsheet/database programs
  • Prevents cheating—exception report will inform if worker did not test once or twice per day
  • Able to prevent unauthorized entry without proper ID/ESD certification; can be upgraded to open electronically locked doors with pass of test
  • Measures from 100 kilohms to 100 megohms, with an option to 1000 megohms
  • Tests performed at 19 volts—not competitive 9 volts
Order Info
Compatible with Microsoft™ Windows™ 95, 98, ME, NT, and 2000 operating systems. Can use Oracle™, Access™, or Excel™ databases. Powered by 9 volt battery (IEC6F22) or 12 volt DC adapter.
Compliance
Conforms to ESD-S 20-20, European EN, DIN and military standards. NIST and RoHs compliant. CE listed.
Specifications
Includes Combo Tester w/ Footplate, Stand, HID Proximity Reader