Learn More
ACL Inc Staticide ESD Safety Shield, 8 oz
Catalog No. NC1052557
Encompass_Preferred
The 6500 formula, like the 6300, is designed for applications on common plastics such as acrylic, PET, PETG, PVC, and polycarbonate, but it is available in an economical aerosol container. A light spray application is sufficient for excellent electrical performance. Coating cures at ambient temperature. Intended for indoor use, this product is ideal for plastic parts, bins, and shelving. Our tests show electricals will last at least two years.
Catalog No. NC1052557 Supplier ACL Inc Supplier No. 6500
The 6500 formula, like the 6300, is designed for applications on common plastics such as acrylic, PET, PETG, PVC, and polycarbonate, but it is available in an economical aerosol container. A light spray application is sufficient for excellent electrical performance. Coating cures at ambient temperature. Intended for indoor use, this product is ideal for plastic parts, bins, and shelving. Our tests show electricals will last at least two years.