Learn More
ACL Inc Staticide Static Sentry, 12oz, 12/cs
Catalog No. 501037865
Encompass_Preferred
Anti-static coatings, aerosol spray, ACL Staticide, Staticide Static Sentry, for use on flooring, carpeting, plastic, glass, worksurfaces, fabric. 67-63-0. Instantly eliminates static charge in less than 2 seconds. No tribocharging; prevents dust attraction. Humidity independent. Ozone safe. Meets MIL-B-81705, Federal Test Standard 101.
Catalog No. 50-103-7865 Supplier ACL Inc Supplier No. 2006
Anti-static coatings, aerosol spray, ACL Staticide, Staticide Static Sentry, for use on flooring, carpeting, plastic, glass, worksurfaces, fabric. 67-63-0. Instantly eliminates static charge in less than 2 seconds. No tribocharging; prevents dust attraction. Humidity independent. Ozone safe. Meets MIL-B-81705, Federal Test Standard 101.