Learn More
ACL Inc Economy 1M Ohm Heel Grounder
Catalog No. NC1841792
Encompass_Preferred
Heel Grounder with 1 inch form fitting elastic garter attached to a rubber sole for comfort fit
Catalog No. NC1841792 Supplier ACL Inc Supplier No. 8120
Heel Grounder with 1 inch form fitting elastic garter attached to a rubber sole for comfort fit