Learn More
ACL Inc Staticide IPA Clean Swab 50pk
Catalog No. NC1654622
Encompass_Preferred
IPA Clean Swab is a high quality, cleanroom swab for the criticalcleaning of small and hard-to-reach areas. The wide paddle headoffers extraordinary cleaning abilities by featuring an inner layerof foam for absorbency and an outer layer of micro-denier forholding contaminants.
Catalog No. NC1654622 Supplier ACL Inc Supplier No. 8020
IPA Clean Swab is a high quality, cleanroom swab for the criticalcleaning of small and hard-to-reach areas. The wide paddle headoffers extraordinary cleaning abilities by featuring an inner layerof foam for absorbency and an outer layer of micro-denier forholding contaminants.