Learn More
ACL Inc General Purpose Staticide, 1 quart, 12/cs
Catalog No. 501037952
Encompass_Preferred
Anti-static coatings, topical anti-stat, ACL Staticide, General Purpose Staticide, quart, anti-static liquid for use on non-porous surfaces such as glass and plastic. 67-63-0.
Catalog No. 50-103-7952 Supplier ACL Inc Supplier No. 2003
Anti-static coatings, topical anti-stat, ACL Staticide, General Purpose Staticide, quart, anti-static liquid for use on non-porous surfaces such as glass and plastic. 67-63-0.