Learn More
Ted Pella Inc Staticide small wipes, 24/box
SDP
Catalog No. NC1262143
Encompass
Staticide is an easy to use topical antistatic solution which can be sprayed on virtually any surface to reduce static charging. It is easily applied on work surfaces, lab benches, chairs, walls, floors, tools, clothing and other materials. The product is non-staining, but we do advise to test first before applying to sensitive surfaces. The antistatic effect last from weeks to months. It prevents static before it builds up and keeps work areas dust-free. The wipes are particularly useful for keyboards, monitors, instruments and equipment where a spray might interfere with sensitive parts such as lenses or electronics. Although it can eliminate charging on non-conductive samples, we do not recommend this product for use on non-conductive samples for imaging in the SEM.
This item is not returnable. View return policy
Catalog No. NC1262143 Supplier Ted Pella Inc Supplier No. 54692
Staticide is an easy to use topical antistatic solution which can be sprayed on virtually any surface to reduce static charging. It is easily applied on work surfaces, lab benches, chairs, walls, floors, tools, clothing and other materials. The product is non-staining, but we do advise to test first before applying to sensitive surfaces. The antistatic effect last from weeks to months. It prevents static before it builds up and keeps work areas dust-free. The wipes are particularly useful for keyboards, monitors, instruments and equipment where a spray might interfere with sensitive parts such as lenses or electronics. Although it can eliminate charging on non-conductive samples, we do not recommend this product for use on non-conductive samples for imaging in the SEM.