Learn More
ACL Inc Economy Wrist Strap Cord Set
Catalog No. NC1841795
Encompass_Preferred
Adjustable/Flexible elastic wrist band lined with 5 rows of double-stiched silver yarn. Includes lightweight 6 ft coiled cord, 4mm socket-to-banana. Includes alligator clip. Darl Blue.
Catalog No. NC1841795 Supplier ACL Inc Supplier No. 8150
Adjustable/Flexible elastic wrist band lined with 5 rows of double-stiched silver yarn. Includes lightweight 6 ft coiled cord, 4mm socket-to-banana. Includes alligator clip. Darl Blue.