Learn More
NRD LLC STATICMASTER BF1 POSITIONER
SDP
Catalog No. 501089916
Encompass_Preferred
STATICMASTER BF1 POSITIONER - USED WITH 2U500 IONIZER, SOLD SEPARATELY
Catalog No. 50-108-9916 Supplier NRD LLC Supplier No. BF1
STATICMASTER BF1 POSITIONER - USED WITH 2U500 IONIZER, SOLD SEPARATELY