Learn More
Thomas Scientific Staticmaster Ionizing Cartridge
SDP
Supplier:  Thomas Scientific 3620A30
Encompass
Static Eliminator Duostat For neutralization of static charge on balances, optics, films, etc. Strength 500 µCi; effective range approximately 40 mm No state or USNRC-specific license is required to purchase or possess this unit. Effective life span is one year. Spent unit may be mailed to the manufacturer for proper disposal. Unit can be placed in or near the item to be neutralized or can be mounted on 3620A50 (1 each of 3620A30 must be purchased to use 3620A50 Single Unit Positioner, A30 Ionizing Units may be used without positioner). Stand of stainless steel with flexible gooseneck arm and maximum height of 15”. Stainless steel housing 3” x 1” x 1/4” high, with mounting tabs. Instantly solves the problem created by static when weighing filters; increases accuracy and repeatability Just one passage through the unit offers reliable, repeatable weighings Portable and easy positioning, requires a minimum amount of bench space. Staticmaster Ionizing Car
Catalog No. NC9952773