Learn More
Volu Sol Statim Stain, 32 oz / 1 L, Volu-Sol™
SDP
Catalog No. NC9745562 Shop All Volu Sol Products
Encompass_Preferred
Proprietary mixture to rapidly stain blood smears (10-15 seconds) and bone marrow (20-30 seconds).  Yields excellent color intensity and morphological differentiation. Eliminates many of the steps involved in the Wright's staining procedure; it includes the buffer within the stain.  Blood smears take less than a minute from start to finish.  Available in both Wright-Giemsa (Stat Stain) and Wright's (Statim Stain) formulations
Catalog No. NC9745562 Supplier Volu Sol Supplier No. VSM032
Proprietary mixture to rapidly stain blood smears (10-15 seconds) and bone marrow (20-30 seconds).  Yields excellent color intensity and morphological differentiation. Eliminates many of the steps involved in the Wright's staining procedure; it includes the buffer within the stain.  Blood smears take less than a minute from start to finish.  Available in both Wright-Giemsa (Stat Stain) and Wright's (Statim Stain) formulations